Learn More
Cayman Chemical CEP-LysIn-d4 500UG
SDP
Catalog No. 502892511
Encompass_Preferred
An internal standard for the quantification of CEP-Lysine by GC- or LC-MS
Catalog No. 50-289-2511 Supplier Cayman Chemical Supplier No. 9000595500 UG
An internal standard for the quantification of CEP-Lysine by GC- or LC-MS