Learn More
CEP Sorbent Spill Kit
SDP
Catalog No. 19147121
Click to view available options
Capacity (English):
65 gal.
Catalog No. Capacity (English)
19-147-121 65 gal.
1 options
Catalog No. 19-147-121 Supplier CEP Supplier No. KIT343
Specifications
Capacity (English) 65 gal.
Capacity (Metric) 246.05 L
Material Polyethylene
Type Spill Control Kit
For Use With (Application) Hazardous materials
Includes (8) 46 in. PIG™ hazmat socks, (5) 3 in. x 10 ft. PIG™ hazmat socks, (1) 5 in. x 10 ft. PIG™ hazmat socks, 5 PIG™ hazmat pillows, 40 PIG™ hazmat mat pads