Learn More
CEP Storm Sentinel Basin Inserts
SDP
Catalog No. 191202578
Click to view available options
Shape:
Circular
Rectangular
Includes:
Packet
Sediment only
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Shape Includes
19-120-2578 Rectangular Packet
19-120-2574 Circular Sediment only
19-120-2575 Rectangular Sediment only
19-120-2577 Circular Packet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-120-2578 Supplier CEP Supplier No. 1344
Specifications
Certifications/Compliance Stormwater pollution prevention
Material Polypropylene Geotextile
Type Storm Sentinel Catch Basin Insert
For Use With (Application) Catch oil and sediment headed into your storm drains and sewers, protects your storm drains and catch basins
For Use With (Equipment) Basins up to 24 x 36 in.
Includes Packet
Shape Rectangular