Learn More
Aviva Systems Biology Corp CEP55 Antibody - middle region (ARP46271_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46271_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CEP55. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CEP55. Alternate names: C10orf3, FLJ10540, URCC6, CT111ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-59291