Learn More
Medchemexpress LLC Ceperognastat (LY3372689) | 2241514-56-5 | 99.1% | 383.44 g·mol⁻¹ | C16H22FN5O3S | 5 MG
SDP
Catalog No. 5000221513
Encompass_Preferred
Ceperognastat is a low-dose, orally available, central nervous system-penetrant inhibitor of O-GlcNAcase (OGA) used in research on tauopathies, including Alzheimer's disease. Preclinical and clinical studies report high brain OGA enzyme occupancy and pharmacokinetic properties consistent with oral CNS drugs. Supplied for research use only.
  • Orally available and central nervous system-penetrant.
  • Achieves high OGA enzyme occupancy in animal and human brain.
  • Potent and selective O-GlcNAcase inhibition for tauopathy research.
  • High purity suitable for laboratory research (≈99.1%).
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-21513 Supplier Medchemexpress LLC Supplier No. HY1446815MG
Ceperognastat is a low-dose, orally available, central nervous system-penetrant inhibitor of O-GlcNAcase (OGA) used in research on tauopathies, including Alzheimer's disease. Preclinical and clinical studies report high brain OGA enzyme occupancy and pharmacokinetic properties consistent with oral CNS drugs. Supplied for research use only.
  • Orally available and central nervous system-penetrant.
  • Achieves high OGA enzyme occupancy in animal and human brain.
  • Potent and selective O-GlcNAcase inhibition for tauopathy research.
  • High purity suitable for laboratory research (≈99.1%).
  • For research use only; not for human or veterinary use.