Learn More
Cayman Chemical Cephacetrile, Purity: ≥95%, CAS: 10206-21-0, 10 mg
SDP
Catalog No. 502913058
Encompass_Preferred
A cephalosporin antibiotic; active against clinical isolates of S. aureus, S. epidermidis, S. pyogenes, E. coli, and Klebsiella (MICs = 0.64, 0.64, 0.16, 6.25 and 3.12 µg/ml, respectively); active against veterinary isolates of S. aureus from bovine mastitis at 30 µg/disc
Catalog No. 50-291-3058 Supplier Cayman Chemical Supplier No. 3626910 MG
A cephalosporin antibiotic; active against clinical isolates of S. aureus, S. epidermidis, S. pyogenes, E. coli, and Klebsiella (MICs = 0.64, 0.64, 0.16, 6.25 and 3.12 µg/ml, respectively); active against veterinary isolates of S. aureus from bovine mastitis at 30 µg/disc