Learn More
Medchemexpress LLC HY-N4118 1mg Medchemexpress, Cephaeline CAS:483-17-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN41181MG
Encompass_Preferred
Medchemexpress, HY-N4118 1mg Cephaeline CAS:483-17-0 Cephaeline is a phenolic alkaloid in Indian Ipecac roots. Cephaeline exhibits potent inhibition of both Zika virus (ZIKV) and Ebola virus (EBOV) infections. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0423