Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cephaeline dihydrochloride | 5853-29-2 | 25 MG
SDP

Catalog No. 5000421984
Encompass_Preferred

Cephaeline dihydrochloride is a selective CYP2D6 inhibitor with an IC50 of 121 μM. It is intended for research use only.

  • Selective CYP2D6 inhibitor with an IC50 of 121 μM
  • Purity of 98.95%
  • White to off-white solid appearance
  • Soluble in DMSO and H2O

Catalog No. 50-004-21984 Supplier Medchemexpress LLC Supplier No. 25MGHYN2260
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cephaeline dihydrochloride is a selective CYP2D6 inhibitor with an IC50 of 121 μM. It is intended for research use only.

  • Selective CYP2D6 inhibitor with an IC50 of 121 μM
  • Purity of 98.95%
  • White to off-white solid appearance
  • Soluble in DMSO and H2O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.