Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cephaeline dihydrochloride | 5853-29-2 | MFCD01732155 | 99.0% | 539.53 | C28H40Cl2N2O4 | 10 MG
SDP

Catalog No. 5000295660
Encompass_Preferred

Cephaeline dihydrochloride is the dihydrochloride salt of cephaeline, a small-molecule research reagent that acts as a selective inhibitor of cytochrome P450 2D6. It is supplied as a solid, white to off-white powder with high purity and solubility properties suitable for in vitro and selected in vivo formulations.

  • Selective CYP2D6 inhibitor with reported IC50 of 121 μM.
  • High purity suitable for research use (≈99%).
  • Molecular formula C28H40Cl2N2O4; molecular weight 539.53.
  • Solid, white to off-white appearance.
  • Soluble in DMSO and water; demonstrated in defined in vivo formulations.
  • Recommended storage sealed, away from moisture and light; long-term storage in solvent at -80°C.
  • Common applications include enzyme inhibition assays and pharmacology studies.

Catalog No. 50-002-95660 Supplier Medchemexpress LLC Supplier No. HYN226010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cephaeline dihydrochloride is the dihydrochloride salt of cephaeline, a small-molecule research reagent that acts as a selective inhibitor of cytochrome P450 2D6. It is supplied as a solid, white to off-white powder with high purity and solubility properties suitable for in vitro and selected in vivo formulations.

  • Selective CYP2D6 inhibitor with reported IC50 of 121 μM.
  • High purity suitable for research use (≈99%).
  • Molecular formula C28H40Cl2N2O4; molecular weight 539.53.
  • Solid, white to off-white appearance.
  • Soluble in DMSO and water; demonstrated in defined in vivo formulations.
  • Recommended storage sealed, away from moisture and light; long-term storage in solvent at -80°C.
  • Common applications include enzyme inhibition assays and pharmacology studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.