Learn More
TARGETMOL CHEMICALS INC Cephaeline hydrochloride 2MG
SDP
Catalog No. 502876386
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Cephaeline hydrochloride, a phenolic alkaloid found in Indian Ipecac, effectively inhibits the infection of viruses ZIKV and EBOV. Purity 97.81%
Catalog No. 50-287-6386 Supplier TARGETMOL CHEMICALS INC Supplier No. T3S18522MG
Also available in 1 mL, 1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Cephaeline hydrochloride, a phenolic alkaloid found in Indian Ipecac, effectively inhibits the infection of viruses ZIKV and EBOV. Purity 97.81%