Learn More
Medchemexpress LLC Cephalexin-d5 | 2101505-56-8 | 98.96% | 5 MG
SDP
Catalog No. 5000405932
Encompass_Preferred
Cephalexin-d5 is deuterium labeled Cephalexin. Cephalexin is a potent, orally active, first-generation cephalosporin antibiotic. It kills gram-positive and some gram-negative bacteria by disrupting the growth of the bacterial cell wall. Cephalexin monohydrate is used for the research of pneumonia, strep throat, and bacterial endocarditis, among other conditions. Deuterium substitution can impact the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-05932 Supplier Medchemexpress LLC Supplier No. HYB0200S5MG
Cephalexin-d5 is deuterium labeled Cephalexin. Cephalexin is a potent, orally active, first-generation cephalosporin antibiotic. It kills gram-positive and some gram-negative bacteria by disrupting the growth of the bacterial cell wall. Cephalexin monohydrate is used for the research of pneumonia, strep throat, and bacterial endocarditis, among other conditions. Deuterium substitution can impact the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS