Learn More
Medchemexpress LLC Cephalexin-d5 (hydrate) | 93.7% | 370.43 | C16H14D5N3O5S | 1 MG
SDP
Catalog No. 5000284244
Encompass_Preferred
Cephalexin-d5 (hydrate) is a deuterium-labeled form of cephalexin provided as a monohydrate solid for analytical and research use. It is supplied for use as a stable-isotope internal standard or tracer in quantitative analysis and pharmacokinetic investigations using mass spectrometry.
  • Deuterium-labeled monohydrate for use as an internal standard in LC-MS/MS.
  • Suitable for quantitative, pharmacokinetic, and metabolic studies.
  • Provided as a 1 mg solid for analytical preparation.
  • Reported purity 93.7%; verify batch COA for exact value.
  • Chemical formula C16H14D5N3O5S; molecular weight 370.43 for identification.
Catalog No. 50-002-84244 Supplier Medchemexpress LLC Supplier No. HYW6539361MG
Cephalexin-d5 (hydrate) is a deuterium-labeled form of cephalexin provided as a monohydrate solid for analytical and research use. It is supplied for use as a stable-isotope internal standard or tracer in quantitative analysis and pharmacokinetic investigations using mass spectrometry.
  • Deuterium-labeled monohydrate for use as an internal standard in LC-MS/MS.
  • Suitable for quantitative, pharmacokinetic, and metabolic studies.
  • Provided as a 1 mg solid for analytical preparation.
  • Reported purity 93.7%; verify batch COA for exact value.
  • Chemical formula C16H14D5N3O5S; molecular weight 370.43 for identification.