Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cephalexin Hydrate, G-Biosciences
SCI_ED

Catalog No. S973231
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S973231 1000 g
S930067 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S973231 Supplier G-Biosciences Supplier No. RC960
Add to Cart
Add to Cart

Cephalexin is a first generation cephalosporin antibiotic most effective against gram-positive cocci.

It also exhibits efficacy against some gram-negative bacilli. In addition, cephalexin has been used to study the penicillin binding proteins, especially PBP3.

Chemical Identifiers

CAS 23325-78-2 , 15686-71-2
Molecular Weight (g/mol) 365.40 g/mol

Specifications

Technique HPLC, Spectrometry, Calibration, Gas Chromatography, UV/Vis
Certifications/Compliance ISO, ISO 9001, GHS/OSHA, REACH, NIST
Type Drug Standard
Element Multi-element
Industry Type Biotech
Quantity 1000 g
Percent Purity >97%
Grade Research
Chemical Name or Material Cephalexin hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.