Learn More
Medchemexpress LLC Cephalexin hydrochloride | 59695-59-9 | 99.8% | 1 ML
SDP
Catalog No. 5000406998
Encompass_Preferred
Cephalexin hydrochloride is a potent, orally active, new semisynthetic cephalosporin antibiotic with a broad antibacterial spectrum. It demonstrates antibacterial activity against a wide variety of gram-positive and gram-negative bacteria by targeting penicillin-binding proteins (PBPs) to inhibit bacterial cell wall assembly. It is utilized in research for conditions such as pneumonia, strep throat, and bacterial endocarditis.
  • Disrupts polymer peptidoglycan (PG) biogenesis
  • Inhibits a broad spectrum of gram-positive and gram-negative organisms
  • Has antibacterial activity in male Swiss-Webster mice with infected bacterial
Catalog No. 50-004-06998 Supplier Medchemexpress LLC Supplier No. HYB0200A10MM/1ML
Cephalexin hydrochloride is a potent, orally active, new semisynthetic cephalosporin antibiotic with a broad antibacterial spectrum. It demonstrates antibacterial activity against a wide variety of gram-positive and gram-negative bacteria by targeting penicillin-binding proteins (PBPs) to inhibit bacterial cell wall assembly. It is utilized in research for conditions such as pneumonia, strep throat, and bacterial endocarditis.
  • Disrupts polymer peptidoglycan (PG) biogenesis
  • Inhibits a broad spectrum of gram-positive and gram-negative organisms
  • Has antibacterial activity in male Swiss-Webster mice with infected bacterial