Learn More
Medchemexpress LLC Cephalexin hydrochloride | 59695-59-9 | 99.8% | 100 MG
SDP
Catalog No. 5000406997
Encompass_Preferred
Cephalexin hydrochloride is a potent, orally active, semisynthetic cephalosporin antibiotic. It demonstrates a broad antibacterial spectrum against various gram-positive and gram-negative bacteria by targeting penicillin-binding proteins (PBPs) to inhibit bacterial cell wall assembly. It is utilized in research for conditions such as pneumonia, strep throat, and bacterial endocarditis.
  • Potent, orally active, semisynthetic cephalosporin antibiotic
  • Broad antibacterial spectrum against gram-positive and gram-negative bacteria
  • Inhibits bacterial cell wall assembly by targeting penicillin-binding proteins
  • Disrupts polymer peptidoglycan biogenesis at 10 μg/mL
  • Exhibits antibacterial activity in animal models
  • Used in research for pneumonia, strep throat, and bacterial endocarditis
Catalog No. 50-004-06997 Supplier Medchemexpress LLC Supplier No. HYB0200A100MG
Cephalexin hydrochloride is a potent, orally active, semisynthetic cephalosporin antibiotic. It demonstrates a broad antibacterial spectrum against various gram-positive and gram-negative bacteria by targeting penicillin-binding proteins (PBPs) to inhibit bacterial cell wall assembly. It is utilized in research for conditions such as pneumonia, strep throat, and bacterial endocarditis.
  • Potent, orally active, semisynthetic cephalosporin antibiotic
  • Broad antibacterial spectrum against gram-positive and gram-negative bacteria
  • Inhibits bacterial cell wall assembly by targeting penicillin-binding proteins
  • Disrupts polymer peptidoglycan biogenesis at 10 μg/mL
  • Exhibits antibacterial activity in animal models
  • Used in research for pneumonia, strep throat, and bacterial endocarditis