Learn More
Research Products International Corp Cephalexin, Monohydrate, 10 Grams
SDP
Catalog No. 50213252
Encompass_Preferred
Cephalexin Monohydrate is a cephalosporin antibiotic. Cephalexin, as a cephalosporin, inhibits bacterial cell wall synthesis by binding to penicillin-binding proteins (PBPs). This interference disrupts the cross-linking of peptidoglycans in the bacterial cell wall, leading to structural instability and eventual cell lysis. 
Cephalexin is sometimes used as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic. Its inclusion in culture medium allows for the specific growth of bacteria lacking resistance to cephalexin.
Catalog No. 50-213-252 Supplier Research Products International Corp Supplier No. C5900010.0
Cephalexin Monohydrate is a cephalosporin antibiotic. Cephalexin, as a cephalosporin, inhibits bacterial cell wall synthesis by binding to penicillin-binding proteins (PBPs). This interference disrupts the cross-linking of peptidoglycans in the bacterial cell wall, leading to structural instability and eventual cell lysis. 
Cephalexin is sometimes used as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic. Its inclusion in culture medium allows for the specific growth of bacteria lacking resistance to cephalexin.