Learn More
Bioworld Cephalexin Monohydrate, 5 g
Catalog No. 50255718
Encompass_Preferred
Cephalexin Monohydrate Active against Gram-positive bacteria. Moderately active against Gram-negative bacteria. Inhibitor of cell wall synthesis. Cell culture tested.β-lactamase sensitive.A semi-synthetic, cephalosporin antibioticFor in vitro lab use only. Not for human use or consumption.Appearance: SolidMelting Point: 196-198°C (lit.)
Catalog No. 50-255-718 Supplier Bioworld Supplier No. 403100121
Cephalexin Monohydrate Active against Gram-positive bacteria. Moderately active against Gram-negative bacteria. Inhibitor of cell wall synthesis. Cell culture tested.β-lactamase sensitive.A semi-synthetic, cephalosporin antibioticFor in vitro lab use only. Not for human use or consumption.Appearance: SolidMelting Point: 196-198°C (lit.)