Learn More
Medchemexpress LLC HY-B0200B 500mg Medchemexpress, Cephalexin (monohydrate) CAS:23325-78-2 Purity:>98%
SDP
Catalog No. 502082406
Encompass_Preferred
Medchemexpress, HY-B0200B 500mg Cephalexin (monohydrate) CAS:23325-78-2 Cephalexin monohydrate is a potent, orally active and the first-generation cephalosporin antibiotic. Cephalexin monohydrate kills gram-positive and some gram-negative bacteria by disrupting the growth of the bacterial cell wall. Cephalexin monohydrate is used for the research of pneumonia, strep throat, and bacterial endocarditis, et al.
Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2406 Supplier Medchemexpress LLC Supplier No. HYB0200B500MG
Medchemexpress, HY-B0200B 500mg Cephalexin (monohydrate) CAS:23325-78-2 Cephalexin monohydrate is a potent, orally active and the first-generation cephalosporin antibiotic. Cephalexin monohydrate kills gram-positive and some gram-negative bacteria by disrupting the growth of the bacterial cell wall. Cephalexin monohydrate is used for the research of pneumonia, strep throat, and bacterial endocarditis, et al.
Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.