Learn More
Medchemexpress LLC Cephalexin monohydrate (cefalexin hydrate) | 23325-78-2 | 99.6% | 1 ML
SDP
Catalog No. 5000405794
Encompass_Preferred
Cephalexin monohydrate is a potent, orally active semisynthetic cephalosporin antibiotic with a broad antibacterial spectrum. It demonstrates antibacterial activity against a wide variety of gram-positive and gram-negative bacteria, and targets penicillin-binding proteins (PBPs) to inhibit bacterial cell wall assembly. This product is for research use only.
  • Potent, orally active semisynthetic cephalosporin antibiotic
  • Broad antibacterial spectrum
  • Active against gram-positive and gram-negative bacteria
  • Targets penicillin-binding proteins (PBPs)
  • Inhibits bacterial cell wall assembly
  • Used for research of pneumonia, strep throat, and bacterial endocarditis
Catalog No. 50-004-05794 Supplier Medchemexpress LLC Supplier No. HYB0200B10MM/1ML
Cephalexin monohydrate is a potent, orally active semisynthetic cephalosporin antibiotic with a broad antibacterial spectrum. It demonstrates antibacterial activity against a wide variety of gram-positive and gram-negative bacteria, and targets penicillin-binding proteins (PBPs) to inhibit bacterial cell wall assembly. This product is for research use only.
  • Potent, orally active semisynthetic cephalosporin antibiotic
  • Broad antibacterial spectrum
  • Active against gram-positive and gram-negative bacteria
  • Targets penicillin-binding proteins (PBPs)
  • Inhibits bacterial cell wall assembly
  • Used for research of pneumonia, strep throat, and bacterial endocarditis