Learn More
Medchemexpress LLC Cephalexin (Standard) | 15686-71-2 | 99.9% | 1 ML
SDP
Catalog No. 5000406995
Encompass_Preferred
Cephalexin is a potent, orally active semisynthetic cephalosporin antibiotic. It has a broad antibacterial spectrum, demonstrating activity against various gram-positive and gram-negative bacteria by targeting penicillin-binding proteins (PBPs) to inhibit bacterial cell wall assembly. This product is for research use only and is not sold to patients.
  • Potent, orally active semisynthetic cephalosporin
  • Broad antibacterial spectrum
  • Demonstrates activity against gram-positive and gram-negative bacteria
  • Inhibits bacterial cell wall assembly by targeting penicillin-binding proteins (PBPs)
  • 99.91% purity
  • Provided as a 10 mM * 1 mL solution in water
  • Suitable for research on pneumonia, strep throat, and bacterial endocarditis
Catalog No. 50-004-06995 Supplier Medchemexpress LLC Supplier No. HYB020010MM/1ML
Cephalexin is a potent, orally active semisynthetic cephalosporin antibiotic. It has a broad antibacterial spectrum, demonstrating activity against various gram-positive and gram-negative bacteria by targeting penicillin-binding proteins (PBPs) to inhibit bacterial cell wall assembly. This product is for research use only and is not sold to patients.
  • Potent, orally active semisynthetic cephalosporin
  • Broad antibacterial spectrum
  • Demonstrates activity against gram-positive and gram-negative bacteria
  • Inhibits bacterial cell wall assembly by targeting penicillin-binding proteins (PBPs)
  • 99.91% purity
  • Provided as a 10 mM * 1 mL solution in water
  • Suitable for research on pneumonia, strep throat, and bacterial endocarditis