Learn More
Medchemexpress LLC Cephalexin (Standard) | 15686-71-2 | 99.9% | 25 MG
SDP
Catalog No. 5000450522
Encompass_Preferred
Cephalexin (Standard) is an analytical standard of Cephalexin, a potent and orally active semisynthetic cephalosporin antibiotic. It demonstrates a broad antibacterial spectrum against various gram-positive and gram-negative bacteria. This product inhibits bacterial cell wall assembly by targeting penicillin-binding proteins (PBPs).
  • Intended for research and analytical applications
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Used for the research of pneumonia, strep throat, and bacterial endocarditis
Catalog No. 50-004-50522 Supplier Medchemexpress LLC Supplier No. 25MGHYB0200R
Cephalexin (Standard) is an analytical standard of Cephalexin, a potent and orally active semisynthetic cephalosporin antibiotic. It demonstrates a broad antibacterial spectrum against various gram-positive and gram-negative bacteria. This product inhibits bacterial cell wall assembly by targeting penicillin-binding proteins (PBPs).
  • Intended for research and analytical applications
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Used for the research of pneumonia, strep throat, and bacterial endocarditis