Learn More
Medchemexpress LLC Cephalexin (Standard) | 15686-71-2 | 99.99% | 100 MG
SDP
Catalog No. 5000407423
Encompass_Preferred
Cephalexin (Standard) is an analytical standard of Cephalexin intended for research and analytical applications. It is a potent, orally active semisynthetic cephalosporin antibiotic with a broad antibacterial spectrum, effective against a wide variety of gram-positive and gram-negative bacteria. It functions by targeting penicillin-binding proteins (PBPs) to inhibit bacterial cell wall assembly. This compound is commonly used in qualitative, quantitative, and methodological research experiments involving techniques such as HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Potent, orally active semisynthetic cephalosporin antibiotic
  • Broad antibacterial spectrum
  • Targets penicillin-binding proteins (PBPs)
  • Inhibits bacterial cell wall assembly
  • Used in research for pneumonia, strep throat, and bacterial endocarditis
  • Suitable for HPLC, GC, and MS applications
Catalog No. 50-004-07423 Supplier Medchemexpress LLC Supplier No. HYB0200R100MG
Cephalexin (Standard) is an analytical standard of Cephalexin intended for research and analytical applications. It is a potent, orally active semisynthetic cephalosporin antibiotic with a broad antibacterial spectrum, effective against a wide variety of gram-positive and gram-negative bacteria. It functions by targeting penicillin-binding proteins (PBPs) to inhibit bacterial cell wall assembly. This compound is commonly used in qualitative, quantitative, and methodological research experiments involving techniques such as HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Potent, orally active semisynthetic cephalosporin antibiotic
  • Broad antibacterial spectrum
  • Targets penicillin-binding proteins (PBPs)
  • Inhibits bacterial cell wall assembly
  • Used in research for pneumonia, strep throat, and bacterial endocarditis
  • Suitable for HPLC, GC, and MS applications