Learn More
Med Vet International Cephalexin Oral Suspension, 250mg/5mL, 100mL, Mix Berry Flavor, Generic Keflex
Catalog No. 502828329
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
250mg/5ml, 100ml Cephalexin oral suspension is a generic pharmaceutical of Keflex. This product is classified as a cephalosporin antibiotic and should be stored at room temperature away from light and moisture.
Catalog No. 50-282-8329 Supplier Med Vet International Supplier No. RX67877054588
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
250mg/5ml, 100ml Cephalexin oral suspension is a generic pharmaceutical of Keflex. This product is classified as a cephalosporin antibiotic and should be stored at room temperature away from light and moisture.