Learn More
AdipoGen Cephalochromin
SDP
Supplier:  AdipoGen BVT0440M001
Encompass_Preferred
Chemical. CAS 25908-26-3. Formula C28H22O10. MW 518.5. Isolated from Pseudoanguillospora sp. strain 6577. Mycotoxin. Bisnaphtha-gamma-pyrone derivative similar to chaetochromin and ustilaginoidin. Antibiotic. Antibacterial, antifungal and anti-algal compound. Antitumor and cytotoxic compound. Calmodulin-sensitive PDE activity inhibitor. Inhibits NO production by activated macrophages. Selective Fabl bacterial enoyl-acyl carrier protein ACP reductase inhibitor. Inducer of G0/G1 cell cycle arrest and apoptosis in human A549 cells.
Catalog No. 50-256-1927