Learn More
Cayman Chemical Cephalomannine, Purity: ≥98%, CAS: 71610-00-9, 10 mg
SDP
Catalog No. 502939133
Encompass_Preferred
A taxane diterpenoid; inhibits calcium-induced microtubule depolymerization in vitro; increases vincristine accumulation in multidrug-resistant 2780AD ovarian cancer cells
Catalog No. 50-293-9133 Supplier Cayman Chemical Supplier No. 2787810 MG
A taxane diterpenoid; inhibits calcium-induced microtubule depolymerization in vitro; increases vincristine accumulation in multidrug-resistant 2780AD ovarian cancer cells