Learn More
Medchemexpress LLC Cephaloridine hydrate | 102039-86-1 | 99.8% | 5 MG
SDP
Catalog No. 5000409856
Encompass_Preferred
Cephaloridine hydrate is a broad-spectrum antibacterial antibiotic. It has certain dose-related nephrotoxicity. For research use only.
  • Inhibits Staphylococcus aureus penicillin-sensitive and resistant strains, Streptococcus pyogenes, Streptococcus pneumoniae, Corynebacterium diphtheriae, and Clostridium septicum.
  • Exhibits dose-related nephrotoxicity in rabbits and monkeys at higher doses.
Catalog No. 50-004-09856 Supplier Medchemexpress LLC Supplier No. HYB2072A5MG
Cephaloridine hydrate is a broad-spectrum antibacterial antibiotic. It has certain dose-related nephrotoxicity. For research use only.
  • Inhibits Staphylococcus aureus penicillin-sensitive and resistant strains, Streptococcus pyogenes, Streptococcus pneumoniae, Corynebacterium diphtheriae, and Clostridium septicum.
  • Exhibits dose-related nephrotoxicity in rabbits and monkeys at higher doses.