Learn More
Cayman Chemical CephalosporIn CzInc 10mg
SDP
Catalog No. 502949970
Encompass_Preferred
A cephalosporin antibiotic; active against a variety of bacteria, including S. pyogenes, C. diphtheriae, E. coli, and H. influenzae (MICs = 31, 15.6, 125, and 31 UG/ml, respectively), as well as clinical isolates of N. gonorrhoeae (MICs = 1.9-15.6 UG/ml)
Catalog No. 50-294-9970 Supplier Cayman Chemical Supplier No. 3326210 MG
A cephalosporin antibiotic; active against a variety of bacteria, including S. pyogenes, C. diphtheriae, E. coli, and H. influenzae (MICs = 31, 15.6, 125, and 31 UG/ml, respectively), as well as clinical isolates of N. gonorrhoeae (MICs = 1.9-15.6 UG/ml)