Learn More
Cayman Chemical Cephalotaxine, Purity: ≥98%, CAS: 24316-19-6, 100 mg
SDP
Catalog No. 502911621
Encompass_Preferred
An alkaloid with anticancer and antiviral activities; cytotoxic to HL-60, NB4, MOLT-4, K562, Jurkat, and Raji cells (IC50s = 4.91, 16.88, 7.09, 22.59, 5.54, and 18.08 µM, respectively); induces mitochondrial apoptosis, as well as impairs lysosomal acidification and autophagic flux in HL-60 cells from 5-20 µM; reduces infectivity and HBsAg secretion in HBV-infected HepG2 2.2.15 cells from 10-100 µM; reduces viral infectivity in Zika virus-infected Vero cells at 20, 40, and 80 µM
Catalog No. 50-291-1621 Supplier Cayman Chemical Supplier No. 36521100 MG
An alkaloid with anticancer and antiviral activities; cytotoxic to HL-60, NB4, MOLT-4, K562, Jurkat, and Raji cells (IC50s = 4.91, 16.88, 7.09, 22.59, 5.54, and 18.08 µM, respectively); induces mitochondrial apoptosis, as well as impairs lysosomal acidification and autophagic flux in HL-60 cells from 5-20 µM; reduces infectivity and HBsAg secretion in HBV-infected HepG2 2.2.15 cells from 10-100 µM; reduces viral infectivity in Zika virus-infected Vero cells at 20, 40, and 80 µM