Learn More
TARGETMOL CHEMICALS INC Cephalothin 200MG
SDP
Catalog No. 502877357
Encompass_Preferred
Also available in 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Cephalothin is a beta-lactam antibiotic, inhibits class C beta-lactamase AmpC (Ki 0.32 uM). Purity 99.62%
Catalog No. 50-287-7357 Supplier TARGETMOL CHEMICALS INC Supplier No. T5570200MG
Also available in 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Cephalothin is a beta-lactam antibiotic, inhibits class C beta-lactamase AmpC (Ki 0.32 uM). Purity 99.62%