Learn More
Selleck Chemical LLC Cephalothin sodium S4594-100mg
SDP
Catalog No. 503108303
Encompass_Preferred
Cephalothin (Cefalotin Cephalotin) sodium is a semisynthetic beta-lactam first-generation cephalosporin antibiotic with bactericidal activity Cephalothin binds to and inactivates penicillin-binding proteins (PBP) located on the inner membrane of the bacterial cell wall
Catalog No. 50-310-8303 Supplier Selleck Chemical LLC Supplier No. S4594100MG
Cephalothin (Cefalotin Cephalotin) sodium is a semisynthetic beta-lactam first-generation cephalosporin antibiotic with bactericidal activity Cephalothin binds to and inactivates penicillin-binding proteins (PBP) located on the inner membrane of the bacterial cell wall