Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cephapirin Benzathine, G-Biosciences
SCI_ED

Catalog No. S935101
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S935101 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S935101 Supplier G-Biosciences Supplier No. RC961
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cephapirin benzathine is a benzathine salt form of a first generation cephalosporin antibiotic. Cephapirin benzathine has a wide range of activity against gram negative and gram positive bacteria.

It has also been used as an antifungal in animals infected with Candida albicans. Benzathine salt has been shown to prolong the duration of antibiotic activity in cephapirin benzathine.

Chemical Identifiers

Molecular Weight (g/mol) 1087.27

Specifications

Physical Form Powder
Quantity 100 mg
Product Line Gibco
Boiling Point 783.9°C
Percent Purity >95%
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.