Learn More
Medchemexpress LLC HY-A0153A 10mg Medchemexpress, Cephapirin (sodium) CAS:24356-60-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYA0153A10MG
Encompass_Preferred
Medchemexpress, HY-A0153A 10mg Cephapirin (sodium) CAS:24356-60-3 Cephapirin sodium (Cefapirin sodium), a semisynthetic cephalosporin antibiotic, is bactericidal against strains of gram-positive and gram-negative bacteria. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0088