Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cephapirin (sodium) | 24356-60-3 | 95.7% | C17H16N3NaO6S2 | 1 ML
SDP

Catalog No. 5000432943
Encompass_Preferred

Cephapirin sodium is an ephalosporin antibiotic known for its broad-spectrum antimicrobial activity. It is effective against various bacterial strains, including Staphylococcus aureus, S. epidermidis, S. viridans, S. pyogenes, and Diplococcus pneumonia.

  • Appearance: White to off-white solid
  • Purity: 95.74%
  • Molecular weight: 445.45
  • Solubility in water: ≥ 100 mg/mL (224.49 mM)
  • Solubility in DMSO: 33.33 mg/mL (74.82 mM)
  • Effective against Staphylococcus aureus in vivo

Catalog No. 50-004-32943 Supplier Medchemexpress LLC Supplier No. HYA0153A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cephapirin sodium is an ephalosporin antibiotic known for its broad-spectrum antimicrobial activity. It is effective against various bacterial strains, including Staphylococcus aureus, S. epidermidis, S. viridans, S. pyogenes, and Diplococcus pneumonia.

  • Appearance: White to off-white solid
  • Purity: 95.74%
  • Molecular weight: 445.45
  • Solubility in water: ≥ 100 mg/mL (224.49 mM)
  • Solubility in DMSO: 33.33 mg/mL (74.82 mM)
  • Effective against Staphylococcus aureus in vivo

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.