Learn More
Medchemexpress LLC Cepharanthine | 481-49-2 | 50 MG
SDP
Catalog No. 5000419205
Encompass_Preferred
Cepharanthine is a natural product isolated from the plant *Stephania cephalantha Hayata*. It demonstrates a wide range of biological activities, including anti-SARS-CoV-2 effects, the ability to reverse multidrug resistance, and inhibitory effects on specific human liver cytochrome P450 enzymes. It also exhibits antitumor, anti-inflammatory, and antinociceptive properties.
  • Exhibits anti-severe acute respiratory syndrome coronavirus 2 (anti-SARS-CoV-2) activities.
  • Suppresses viral proliferation.
  • Reverses P-gp-mediated multidrug resistance in K562 cells.
  • Enhances the sensitivity of anticancer agents in xenograft mouse models.
  • Shows inhibitory effects on human liver cytochrome P450 enzymes CYP3A4, CYP2E1, and CYP2C9.
  • Possesses antitumor, anti-inflammatory, and antinociceptive effects.
Catalog No. 50-004-19205 Supplier Medchemexpress LLC Supplier No. HYN697250MG
Cepharanthine is a natural product isolated from the plant *Stephania cephalantha Hayata*. It demonstrates a wide range of biological activities, including anti-SARS-CoV-2 effects, the ability to reverse multidrug resistance, and inhibitory effects on specific human liver cytochrome P450 enzymes. It also exhibits antitumor, anti-inflammatory, and antinociceptive properties.
  • Exhibits anti-severe acute respiratory syndrome coronavirus 2 (anti-SARS-CoV-2) activities.
  • Suppresses viral proliferation.
  • Reverses P-gp-mediated multidrug resistance in K562 cells.
  • Enhances the sensitivity of anticancer agents in xenograft mouse models.
  • Shows inhibitory effects on human liver cytochrome P450 enzymes CYP3A4, CYP2E1, and CYP2C9.
  • Possesses antitumor, anti-inflammatory, and antinociceptive effects.