Learn More
Selleck Chemical LLC Cepharanthine-50mg
SDP
Catalog No. 502389598
Encompass_Preferred
Cepharanthine (NSC-623442) is a biscoclaurine alkaloid inhibiting tumor necrosis factor (TNF)-α-mediated NFκB stimulation, plasma membrane lipid peroxidation and platelet aggregation and suppressing cytokine production.
Catalog No. 50-238-9598 Supplier Selleck Chemical LLC Supplier No. S423850MG
Cepharanthine (NSC-623442) is a biscoclaurine alkaloid inhibiting tumor necrosis factor (TNF)-α-mediated NFκB stimulation, plasma membrane lipid peroxidation and platelet aggregation and suppressing cytokine production.