Learn More
Cayman Chemical Cepharanthine, Purity: ≥98%, CAS: 481-49-2, 100 mg
SDP
Catalog No. 502984300
Encompass_Preferred
A biscoclaurine alkaloid that can decrease the fluidity of biological membranes; exhibits antiparasitic, antimalarial, antiviral, anti-inflammatory, antimetastatic, and anticancer activities in various cell and animal models
Catalog No. 50-298-4300 Supplier Cayman Chemical Supplier No. 19648100 MG
A biscoclaurine alkaloid that can decrease the fluidity of biological membranes; exhibits antiparasitic, antimalarial, antiviral, anti-inflammatory, antimetastatic, and anticancer activities in various cell and animal models