Learn More
Cayman Chemical Cepharanthine, Purity: ≥98%, CAS: 481-49-2, 50 mg
SDP
Catalog No. 502981337
Encompass_Preferred
A biscoclaurine alkaloid that can decrease the fluidity of biological membranes; exhibits antiparasitic, antimalarial, antiviral, anti-inflammatory, antimetastatic, and anticancer activities in various cell and animal models
Catalog No. 50-298-1337 Supplier Cayman Chemical Supplier No. 1964850 MG
A biscoclaurine alkaloid that can decrease the fluidity of biological membranes; exhibits antiparasitic, antimalarial, antiviral, anti-inflammatory, antimetastatic, and anticancer activities in various cell and animal models