Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MicroBiologics™ Cepheid Xpert™ Carba-R Control Panel
SDP

Catalog No. 23017286 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
21 Pellets of 8 Organisms
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-017-286 21 Pellets of 8 Organisms
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-017-286 Supplier MicroBiologics™ Supplier No. 8187
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Six positive control swabs, Six negative control swabs and certificate of analysis

Features manufacturers recommended QC strains in an easy-to-use format.

  • Intact pathogens inactivated by chemical, radiological, or heat treatments

Organisms

Positive Control:

  • Klebsiella pneumoniae derived from NCTC 13440
  • Klebsiella pneumoniae derived from NCTC 13443
  • Klebsiella pneumoniae derived from NCTC 13438
  • Klebsiella pneumoniae derived from NCTC 13442

Negative Control

  • Escherichia coli derived from NCTC 11954

Specifications

Content And Storage 2 to 25°C
Format Helix Elite™ single use foil pouch
Product Type Lyophilized Organism
Target Organism Class Enteric Bacteria
Test Type Verification Panel
For Use With (Application) Used as positive control
For Use With (Equipment) Cepheid Xpert™ assay kits
Quantity 21 Pellets of 8 Organisms
Specimen Type Inactivated Lyophilized Microorganisms

† The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark and the ATCC catalog marks are trademarks of ATCC. Microbiologics, Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.