Learn More
Microbiologics™ Helix Elite™ Cepheid Xpert TV Control Panel
SDP
Catalog No. 23004139 Shop All Microbiologics Products
Change view
Click to view available options
Six positive and negative control swabs
Catalog No.
23-004-139 Six positive and negative control swabs
1 options
This item is not returnable. View return policy
Catalog No. 23-004-139 Supplier Microbiologics™ Supplier No. 8189
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
Six positive control swabs and six negative control swabs. The positive control consists of Trichomonas vaginalis and human cells. The negative control consists of Neisseria gonorrhoeae and human cells.
Features the manufacturer recommended QC strains in an easy-to-use format
Specifications
Biosafety Level 1
Description Helix Elite Cepheid Xpert TV Control Panel
Detects Trichomonas vaginalis