Learn More
Medchemexpress LLC Cephradine monohydrate | 75975-70-1 | 91.0% | 100 MG
SDP
Catalog No. 5000342582
Encompass_Preferred
Cephradine monohydrate is a broad-spectrum, orally active cephalosporin effective against both gram-positive and gram-negative pathogens. It eradicates most penicillinase-producing organisms resistant to penicillin G, penicillin V, and ampicillin. This compound has been utilized in research concerning genitourinary, gastrointestinal, and respiratory tract infections, as well as infections of the skin and soft tissues. It also blocks solar-ultraviolet induced skin inflammation.
  • Active against gram-positive and gram-negative pathogens.
  • Effective in eradicating most penicillinase-producing organisms resistant to penicillin G, penicillin V, and ampicillin.
  • Used in research of genitourinary, gastrointestinal, and respiratory tract infections.
  • Used in research of infections of the skin and soft tissues.
  • Blocks solar-ultraviolet induced skin inflammation through direct inhibition of TOPK.
  • Targets β-lactam.
Catalog No. 50-003-42582 Supplier Medchemexpress LLC Supplier No. HY128449100MG
Cephradine monohydrate is a broad-spectrum, orally active cephalosporin effective against both gram-positive and gram-negative pathogens. It eradicates most penicillinase-producing organisms resistant to penicillin G, penicillin V, and ampicillin. This compound has been utilized in research concerning genitourinary, gastrointestinal, and respiratory tract infections, as well as infections of the skin and soft tissues. It also blocks solar-ultraviolet induced skin inflammation.
  • Active against gram-positive and gram-negative pathogens.
  • Effective in eradicating most penicillinase-producing organisms resistant to penicillin G, penicillin V, and ampicillin.
  • Used in research of genitourinary, gastrointestinal, and respiratory tract infections.
  • Used in research of infections of the skin and soft tissues.
  • Blocks solar-ultraviolet induced skin inflammation through direct inhibition of TOPK.
  • Targets β-lactam.