Learn More
Apexbio Technology LLC CEPT Cocktail, 1 kit, A human pluripotent stem cell (hPSCs) culture supplement for stem cell research
SDP
Catalog No. NC2949335
Encompass_Preferred
CEPT Cocktail is a combination of small molecules developed by the National Institutes of Health for use in stem cell research. CEPT Cocktail consists of four compounds, namely Chroman 1, Emricasan, Trans-ISRIB, and Polyamine Supplement. Studies have shown that CEPT can significantly prevent cell stress and DNA damage of human pluripotent stem cells (hPSCs) during passaging, and can improve the survival rate of hPSCs during passaging and cryopreservation. In addition, CEPT can also improve cell survival in a variety of stem cell studies, such as embryo-like and organoid formation, single-cell cloning, and gene editing.
Catalog No. NC2949335 Supplier Apexbio Technology LLC Supplier No. K27141
CEPT Cocktail is a combination of small molecules developed by the National Institutes of Health for use in stem cell research. CEPT Cocktail consists of four compounds, namely Chroman 1, Emricasan, Trans-ISRIB, and Polyamine Supplement. Studies have shown that CEPT can significantly prevent cell stress and DNA damage of human pluripotent stem cells (hPSCs) during passaging, and can improve the survival rate of hPSCs during passaging and cryopreservation. In addition, CEPT can also improve cell survival in a variety of stem cell studies, such as embryo-like and organoid formation, single-cell cloning, and gene editing.