Learn More
Medchemexpress LLC CER10-d9 | 2260669-52-9 | 99.0% | 548.97 | 1 MG
SDP
Catalog No. 5000360677
Encompass_Preferred
CER10-d9 is a deuterium-labeled version of CER10, designed for research purposes. It is explicitly stated that it is not for sale to patients. This compound is offered in solid form and presents as a white to off-white substance.
  • Tracer application
  • Internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS
  • Deuterium labeling incorporates stable heavy isotopes of hydrogen
  • Availability in multiple quantities
  • Quality boasts a purity of 99.0%
Catalog No. 50-003-60677 Supplier Medchemexpress LLC Supplier No. HY146828S1MG
CER10-d9 is a deuterium-labeled version of CER10, designed for research purposes. It is explicitly stated that it is not for sale to patients. This compound is offered in solid form and presents as a white to off-white substance.
  • Tracer application
  • Internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS
  • Deuterium labeling incorporates stable heavy isotopes of hydrogen
  • Availability in multiple quantities
  • Quality boasts a purity of 99.0%