Learn More
Avanti Polar Lipids CER2(D9) 5MG
Supplier:  Avanti Polar Lipids 791660P5MG
Encompass
N-palmitoyl(d9) D-erythro-sphingosine 5mg | Purity: >99% | CAS Number 2260669-51-8 | Formula Weight: 546.956
Catalog No. NC2004367