Learn More
Medchemexpress LLC CER3-d9 | 2260669-53-0 | 99% | 564.97 | 1 MG
SDP
Catalog No. 5000362550
Encompass_Preferred
CER3-d9 is a deuterium-labeled version of CER3. It is intended for research use only. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration, the process of incorporating stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules, has shown potential in affecting the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-62550 Supplier Medchemexpress LLC Supplier No. HY146831S1MG
CER3-d9 is a deuterium-labeled version of CER3. It is intended for research use only. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration, the process of incorporating stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules, has shown potential in affecting the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS