Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cer5-2'R-d9 | 2260670-31-1 | 99.0% | 562.96 | 1 MG
SDP

Catalog No. 5000360725
Encompass_Preferred

CER5-2'R-d9 is a deuterium-labeled CER5-2'R.

  • Can serve as a tracer.
  • Can be used as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Incorporation of stable heavy isotopes into drug molecules for quantitation during drug development.
  • Deuteration can influence the pharmacokinetic and metabolic profiles of drugs.

Catalog No. 50-003-60725 Supplier Medchemexpress LLC Supplier No. HY146909S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

CER5-2'R-d9 is a deuterium-labeled CER5-2'R.

  • Can serve as a tracer.
  • Can be used as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Incorporation of stable heavy isotopes into drug molecules for quantitation during drug development.
  • Deuteration can influence the pharmacokinetic and metabolic profiles of drugs.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.