Learn More
Avanti Polar Lipids CER5-2'S(D9) 1MG
Catalog No. NC2004354
Encompass
N-(2'-(S)-hydroxypalmitoyl(d9)) D-erythro-sphingosine 1mg | Purity>99% | CAS 2260670-30-0 | Formula Weight - 562.956
Catalog No. NC2004354 Supplier Avanti Polar Lipids Supplier No. 860847P1MG
N-(2'-(S)-hydroxypalmitoyl(d9)) D-erythro-sphingosine 1mg | Purity>99% | CAS 2260670-30-0 | Formula Weight - 562.956