Learn More
Medchemexpress LLC Cer6-2'r-d9 | 2260670-10-6 | 99.0% | 580.97 | 1 MG
SDP
Catalog No. 5000363009
Encompass_Preferred
CER6-2'R-d9 is a deuterium-labeled variant of CER6-2'R, designed for research applications. It functions as a tracer or an internal standard, facilitating quantitative analysis across various advanced spectroscopic techniques.
  • Deuterium-labeled compound
  • Acts as a tracer for research
  • Serves as an internal standard for quantitative analysis
  • Compatible with NMR, GC-MS, and LC-MS analytical methods
  • Intended for research use only
Catalog No. 50-003-63009 Supplier Medchemexpress LLC Supplier No. HY146899S1MG
CER6-2'R-d9 is a deuterium-labeled variant of CER6-2'R, designed for research applications. It functions as a tracer or an internal standard, facilitating quantitative analysis across various advanced spectroscopic techniques.
  • Deuterium-labeled compound
  • Acts as a tracer for research
  • Serves as an internal standard for quantitative analysis
  • Compatible with NMR, GC-MS, and LC-MS analytical methods
  • Intended for research use only