Learn More
Medchemexpress LLC CER6-2'S-d9 | 2260670-11-7 | 99.0% | 580.97 | 1 MG
SDP
Catalog No. 5000363558
Encompass_Preferred
CER6-2'S-d9 is deuterium labeled CER6-2'S, intended for research use only. This compound serves as a valuable tool in drug development, where stable heavy isotopes are incorporated into drug molecules, primarily as tracers for quantitative analysis. Deuteration, in particular, is noted for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-63558 Supplier Medchemexpress LLC Supplier No. HY146905S1MG
CER6-2'S-d9 is deuterium labeled CER6-2'S, intended for research use only. This compound serves as a valuable tool in drug development, where stable heavy isotopes are incorporated into drug molecules, primarily as tracers for quantitative analysis. Deuteration, in particular, is noted for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS