Learn More
Medchemexpress LLC HY-19323 10mg Medchemexpress, Ceralasertib CAS:1352226-88-0 Purity:>98%
SDP
Catalog No. 502029276
Encompass_Preferred
Medchemexpress, HY-19323 10mg Ceralasertib CAS:1352226-88-0 Ceralasertib (AZD6738) is an orally active and bioavailable inhibitor of ATR kinase with an IC50 of 1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9276 Supplier Medchemexpress LLC Supplier No. HY1932310MG
Medchemexpress, HY-19323 10mg Ceralasertib CAS:1352226-88-0 Ceralasertib (AZD6738) is an orally active and bioavailable inhibitor of ATR kinase with an IC50 of 1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.