Learn More
Cayman Chemical Ceralifimod, Purity: ≥95%, CAS: 891859-12-4, 10 mg
SDP
Catalog No. 502936892
Encompass_Preferred
An agonist of S1P1 and S1P5 receptors (Kis = 0.626 and 0.574 nM, respectively); selective for S1P1 and S1P5 over S1P2-4 (Kis = >5,450, >5,630, and 28.8 nM, respectively); induces calcium influx in CHO-K1 cells overexpressing S1P1 (EC50 = 1 nM) and cAMP accumulation in CHO-K1 cells overexpressing S1P1 or S1P5 (EC50s = 0.027 and 0.33 nM, respectively); reduces paw edema in a rat model of CIA at 0.03 and 0.1 mg/kg; delays disease onset and inhibits lymphocyte infiltration of the spinal cord in a rat model of EAE; prevents disease relapse in a non-obese diabetic mouse model of relapsing-remitting EAE
Catalog No. 50-293-6892 Supplier Cayman Chemical Supplier No. 2415910 MG
An agonist of S1P1 and S1P5 receptors (Kis = 0.626 and 0.574 nM, respectively); selective for S1P1 and S1P5 over S1P2-4 (Kis = >5,450, >5,630, and 28.8 nM, respectively); induces calcium influx in CHO-K1 cells overexpressing S1P1 (EC50 = 1 nM) and cAMP accumulation in CHO-K1 cells overexpressing S1P1 or S1P5 (EC50s = 0.027 and 0.33 nM, respectively); reduces paw edema in a rat model of CIA at 0.03 and 0.1 mg/kg; delays disease onset and inhibits lymphocyte infiltration of the spinal cord in a rat model of EAE; prevents disease relapse in a non-obese diabetic mouse model of relapsing-remitting EAE